Innovatek Cytology Brushes
Innovatek Cytology Brushes for use with PCR and Sample Collection
$494.89 - $494.89
Specifications
Product Type | Cytology Brush |
---|---|
Sterility | Non-sterile |
Length (English) Handle | 8 in. |
For Use With (Application) | Pap Smears, PCR, STD Sample Collection |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
36-102-6743
|
Innovatek
4201CB8B |
1000 Brushes/Cs. |
Case of 1000 Each for $494.89
|
|
|||||
Description
- For Pap smears and sexually transmitted disease sample collection
- Can be used for PCR testing
- Soft nylon brush
Specifications
Cytology Brush | |
Non-sterile | |
8 in. | |
Pap Smears, PCR, STD Sample Collection |