Learn More
Promega CytoTox-Fluor™ Cytotoxicity Assay
The CytoTox-Fluor Cytotoxicity Assay is a single-reagent-addition, homogeneous, fluorescent assay that measures the relative number of dead cells in cell populations.
$262.20 - $1,843.32
Description
- A Fluorescent Assay to Measure the Relative Number of Dead Cells in Culture
- Single-reagent-addition, homogeneous assay
- Amenable to multiplexing with other luminescent cell-based assays
Specifications
| Fluorescence |
Patent pending