Promega CytoTox-Glo™ Cytotoxicity Assay DFS Item
The CytoTox-Glo™ Assay is a luminescent cytotoxicity assay that measures the relative number of dead cells in a population.
Supplier:  Promega G9292
 View more versions of this product
Catalog No. PRG9292
Description
Description
  • Measures Cytotoxicity Due to Loss of Membrane Integrity
  • Measures extracellular activity of dead-cell protease
  • Easy and fast add-mix-measure protocol
  • Powerful luminescent detection
Specifications
Specifications
2 x 50 mL
-30°C to -10°C