missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ CytoVista™ Tissue Clearing/Staining Kit DFS Item

CytoVista™ Tissue Clearing/Staining Kit clears fluorescently labeled tissue to enable sharp and bright 3D fluorescent imaging at any tissue depth.

Supplier:  Invitrogen™ V11324

Catalog No. V11324


Explore available promotions

Description

Description

Kit contains both CytoVista™ Tissue Clearing Reagent and CytoVista™ Tissue Clearing Enhancer, which together can be used to clear most fluorescently labeled tissue types. The kit also contains buffers for immunohistochemistry (IHC) of tissue.

Features of the CytoVista™ family of tissue and 3D cell culture clearing reagents include:
• Rapid clearing of fluorescently labeled tissue for 3D imaging
• Minimal shrinkage, expansion, or other morphological changes to tissue
• Simple workflow does not require any special instruments or equipment and is easily adapted to IHC
• Compatible with most fluorophores, including dyes and fluorescent proteins

The CytoVista™ tissue and 3D cell culture clearing workflow clears tissue or 3D cultured cells with minimal changes to morphology or the sensitivity of detection with almost any fluorophore. With an easy-to-use protocol, whole mouse brain (8-mm section) can be cleared in 24 hours, or a 1-mm section in 2 hours, without the use of any special instruments or equipment.

The CytoVista™ clearing workflow is compatible with most fluorophores, including fluorescent proteins that are detected with common fluorescent imaging instruments, such as wide-field microscopes, high content analyzers, confocal microscopes, and lightsheet microscopes. For first time users, we recommend the CytoVista™ Tissue Clearing/Staining Kit (Cat No V11324) or CytoVista™ 3D Cell Culture Clearing/Staining Kit (Cat No. V11325). These kits contain all necessary reagents for both clearing and IHC staining.

CytoVista™ clearing technology sufficiently clears tissue for 3D fluorescent imaging, but unlike other clearing techniques does not significantly changes the tissue morphology. Minimal morphological changes, such as shrinkage and contraction, have been observed. For precious samples, the clearing can be reversed, and the tissue can be processed for histology studies, like H&E staining.

For tissues that are easier to clear, like brain, lung, intestine, skin, and tissues of less than 250-μm thickness, treatment with CytoVista™ Tissue Clearing Reagent is sufficient for 3D imaging. However, tissues of more than 250-μm thickness or that are hard to clear like kidney, liver, heart, and placenta will require an initial treatment with CytoVista™ Tissue Clearing Reagent followed by treatment with CytoVista™ Tissue Clearing Enhancer.

CytoVista™ 3D Cell Culture Clearing Reagent is recommended for spheroids or organoids up to 1 mm in thickness.

For hard tissues like bone, cartilage, and connective tissue, CytoVista™ Tissue Clearing Reagent is not recommended.

Specifications

Specifications

Tissue Clearing/Staining Kit
Cell & Tissue Preparation Reagent
1 kit
CytoVista
  • CytoVista Tissue Clearing Reagent, 30 mL, store at room temperature
  • CytoVista Tissue Clearing Enhancer, 30 mL, store at room temperature
  • CytoVista Antibody Penetration Buffer, 30 mL, store at 2°C to 8°CC
  • CytoVista 10X Wash Buffer, 30 mL, store at 2°C to 8°CC
  • CytoVista Blocking Buffer, 30 mL, store at 2°C to 8°CC
  • CytoVista Antibody Dilution Buffer, 30 mL, store at 2°C to 8°CC
  • CytoVista Permeabilization Buffer, 30 mL, store at 2°C to 8°CC

    Due to variable shipping temperatures, content may be shipped in mulitple containers.
Approved for shipment at Room Temperature or on Wet Ice
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.