missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals D-Alanine, 99%
Supplier: Thermo Scientific Chemicals A1023114
Description
D-Alanine is essential for the biosynthesis of peptidoglycan crosslinking sub-units that are used for bacterial cell walls.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Alanine is essential for the biosynthesis of peptidoglycan crosslinking sub-units that are used for bacterial cell walls.
Solubility
Soluble in water (155 mg/ml at 20°C), ethanol (slightly), and pyridine (slightly). Insoluble in ether, and acetone.
Notes
Store in cool, dry conditions in well sealed containers. Strong oxidizing agents.
Specifications
∼295°C (decomposition) | |
338-69-2 | |
MFCD00008077 | |
14,204 | |
Soluble in water (155mg/ml at 20°C),ethanol (slightly),and pyridine (slightly). Insoluble in ether,and acetone. | |
C[C@@H](N)C(O)=O | |
25 g | |
71080 | |
−14° (c=5 in 5N HCl) | |
99% |
D-Alanine | |
C3H7NO2 | |
1720249 | |
d-alanine, 2r-2-aminopropanoic acid, r-alanine, h-d-ala-oh, d-2-aminopropionic acid, d-ala, r-2-aminopropanoic acid, d-alpha-alanine, alanine, d, alanine d-form | |
QNAYBMKLOCPYGJ-UWTATZPHSA-N | |
(2R)-2-aminopropanoic acid | |
89.09 | |
CHEBI:15570 | |
89.09 |
Chemical Identifiers
338-69-2 | |
89.09 | |
QNAYBMKLOCPYGJ-UWTATZPHSA-N | |
71080 | |
(2R)-2-aminopropanoic acid |
C3H7NO2 | |
MFCD00008077 | |
d-alanine, 2r-2-aminopropanoic acid, r-alanine, h-d-ala-oh, d-2-aminopropionic acid, d-ala, r-2-aminopropanoic acid, d-alpha-alanine, alanine, d, alanine d-form | |
CHEBI:15570 | |
C[C@@H](N)C(O)=O |
Safety and Handling
EINECSNumber : 206-418-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only