missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals D-Alanine, 99%

Supplier:  Thermo Scientific Chemicals A1023122

 View more versions of this product

Catalog No. AAA1023122


Explore available promotions

Description

Description

D-Alanine is essential for the biosynthesis of peptidoglycan crosslinking sub-units that are used for bacterial cell walls.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Alanine is essential for the biosynthesis of peptidoglycan crosslinking sub-units that are used for bacterial cell walls.

Solubility
Soluble in water (155 mg/ml at 20°C), ethanol (slightly), and pyridine (slightly). Insoluble in ether, and acetone.

Notes
Store in cool, dry conditions in well sealed containers. Strong oxidizing agents.
Specifications

Specifications

∼295°C (decomposition)
338-69-2
MFCD00008077
14,204
Soluble in water (155mg/ml at 20°C),ethanol (slightly),and pyridine (slightly). Insoluble in ether,and acetone.
C[C@@H](N)C(O)=O
100 g
71080
−14° (c=5 in 5N HCl)
99%
D-Alanine
C3H7NO2
1720249
d-alanine, 2r-2-aminopropanoic acid, r-alanine, h-d-ala-oh, d-2-aminopropionic acid, d-ala, r-2-aminopropanoic acid, d-alpha-alanine, alanine, d, alanine d-form
QNAYBMKLOCPYGJ-UWTATZPHSA-N
(2R)-2-aminopropanoic acid
89.09
CHEBI:15570
89.09

Chemical Identifiers

338-69-2
89.09
QNAYBMKLOCPYGJ-UWTATZPHSA-N
71080
(2R)-2-aminopropanoic acid
C3H7NO2
MFCD00008077
d-alanine, 2r-2-aminopropanoic acid, r-alanine, h-d-ala-oh, d-2-aminopropionic acid, d-ala, r-2-aminopropanoic acid, d-alpha-alanine, alanine, d, alanine d-form
CHEBI:15570
C[C@@H](N)C(O)=O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 206-418-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only