missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals D-Alanine, +99%
$84.05 - $280.77
Chemical Identifiers
CAS | 338-69-2 |
---|---|
Molecular Formula | C3H7NO2 |
Molecular Weight (g/mol) | 89.09 |
MDL Number | MFCD00008077 |
InChI Key | QNAYBMKLOCPYGJ-UWTATZPHSA-N |
Synonym | d-alanine, 2r-2-aminopropanoic acid, r-alanine, h-d-ala-oh, d-2-aminopropionic acid, d-ala, r-2-aminopropanoic acid, d-alpha-alanine, alanine, d, alanine d-form |
PubChem CID | 71080 |
ChEBI | CHEBI:15570 |
IUPAC Name | (2R)-2-aminopropanoic acid |
SMILES | C[C@@H](N)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC172080050
|
Thermo Scientific Chemicals
172080050 |
5 g | Glass Bottle |
Each for $84.05
|
|
||||
AC172080250
|
Thermo Scientific Chemicals
172080250 |
25 g | Glass Bottle |
Each for $280.77
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
338-69-2 | |
89.09 | |
QNAYBMKLOCPYGJ-UWTATZPHSA-N | |
71080 | |
(2R)-2-aminopropanoic acid |
C3H7NO2 | |
MFCD00008077 | |
d-alanine, 2r-2-aminopropanoic acid, r-alanine, h-d-ala-oh, d-2-aminopropionic acid, d-ala, r-2-aminopropanoic acid, d-alpha-alanine, alanine, d, alanine d-form | |
CHEBI:15570 | |
C[C@@H](N)C(O)=O |
Specifications
278.0°C to 282.0°C | |
2ppm max. (As2O3) | |
0.1% max. | |
Authentic | |
99+% | |
Glass Bottle | |
MFCD00008077 | |
95% min. (c=10, H2O, 430nm) 1cm cell | |
d-alanine, 2r-2-aminopropanoic acid, r-alanine, h-d-ala-oh, d-2-aminopropionic acid, d-ala, r-2-aminopropanoic acid, d-alpha-alanine, alanine, d, alanine d-form | |
QNAYBMKLOCPYGJ-UWTATZPHSA-N | |
(2R)-2-aminopropanoic acid | |
5 g | |
CHEBI:15570 | |
≥99% | |
D-Alanine |
338-69-2 | |
White | |
10ppm max. | |
0.2% max. (105°C, 3 hrs) | |
C3H7NO2 | |
CH3CH(NH2)CO2H | |
− 14.50 | |
0.2% max. | |
Solubility in water: 155g/L in water (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and organic solvents | |
C[C@@H](N)C(O)=O | |
89.09 | |
71080 | |
89.09 | |
Crystalline Powder or Crystals |
RUO – Research Use Only