missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-alpha-Tocopherol Succinate 97.0+%, TCI America™

Supplier:  TCI America T26285G

 View more versions of this product

Catalog No. T26285G


Explore available promotions

Description

Description

Specifications

Specifications

D-alpha-Tocopherol Succinate
75°C
C33H54O5
5 g
IELOKBJPULMYRW-NJQVLOCASA-N
4-oxo-4-[[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl]oxy]butanoic acid
20353
≥97.0% (GC,T)
4345-03-3
White-Yellow
MFCD00072055
Vitamin E Succinate
CC1=C2C(=C(C(=C1C)OC(=O)CCC(=O)O)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C
530.79
530.79
Crystalline Powder

Chemical Identifiers

4345-03-3
530.79
IELOKBJPULMYRW-NJQVLOCASA-N
20353
CC1=C2C(=C(C(=C1C)OC(=O)CCC(=O)O)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C
C33H54O5
MFCD00072055
Vitamin E Succinate
4-oxo-4-[[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl]oxy]butanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0477

RTECSNumber : EJ9984000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions