missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals D-(-)-Arabinose, 99%

Supplier:  Thermo Scientific Chemicals A1035709

 View more versions of this product

Catalog No. AAA1035709


Explore available promotions

Description

Description

D-(-)-Arabinose is used in culture media for some bacteria. It serves as a component of biopolymers such as hemicellulose and pectin. In the synthetic biology laboratory, it is employed as a reversible switch for protein expression under the Pbad promoter (i.e. part of plasmids) in E. coli. Further, it acts as an inhibitor of the enzyme glucose dehydrogenase. In addition to this, it is used as a pharmaceutical intermediate and food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-Arabinose is used in culture media for some bacteria. It serves as a component of biopolymers such as hemicellulose and pectin. In the synthetic biology laboratory, it is employed as a reversible switch for protein expression under the Pbad promoter (i.e. part of plasmids) in E. coli. Further, it acts as an inhibitor of the enzyme glucose dehydrogenase. In addition to this, it is used as a pharmaceutical intermediate and food additive.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Specifications

D- (-)-Arabinose
154°C to 158°C
MFCD00135608
1723079
Soluble in water.
−104° (c=10 in water, 24h)
(2S,3R,4R)-2,3,4,5-tetrahydroxypentanal
66308
150.13
10323-20-3
C5H10O5
10 g
d---arabinose, d-arabinose, dl-arabinose, arabinose, d, arabinose,d, 2s,3r,4r-2,3,4,5-tetrahydroxypentanal, unii-f0w6etz4e5, --arabinose, aldehydo-d-arabinose, f0w6etz4e5
PYMYPHUHKUWMLA-WDCZJNDASA-N
C(C(C(C(C=O)O)O)O)O
150.13
CHEBI:46983
99%

Chemical Identifiers

10323-20-3
150.13
PYMYPHUHKUWMLA-WDCZJNDASA-N
66308
(2S,3R,4R)-2,3,4,5-tetrahydroxypentanal
C5H10O5
MFCD00135608
d---arabinose, d-arabinose, dl-arabinose, arabinose, d, arabinose,d, 2s,3r,4r-2,3,4,5-tetrahydroxypentanal, unii-f0w6etz4e5, --arabinose, aldehydo-d-arabinose, f0w6etz4e5
CHEBI:46983
C(C(C(C(C=O)O)O)O)O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 233-708-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only