missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Cycloserine, 20mg/mL in distilled water, sterile-filtered, Thermo Scientific™
$271.10 - $490.84
Chemical Identifiers
CAS | 68-41-7 |
---|---|
Molecular Formula | C3H6N2O2 |
Molecular Weight (g/mol) | 102.093 |
MDL Number | MFCD00005353 |
InChI Key | DYDCUQKUCUHJBH-UWTATZPHSA-N |
Synonym | d-cycloserine, cycloserine, seromycin, orientomycin, oxamycin, cyclorin, cyclo-d-serine, cicloserina, farmiserina, miroseryn |
PubChem CID | 6234 |
ChEBI | CHEBI:40009 |
IUPAC Name | (4R)-4-amino-1,2-oxazolidin-3-one |
SMILES | C1C(C(=O)NO1)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ67020AC
|
Thermo Scientific Chemicals
J67020AC |
25 mL |
Each for $271.10
|
|
|||||
AAJ67020AD
|
Thermo Scientific Chemicals
J67020AD |
50 mL |
Each for $490.84
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
68-41-7 | |
102.093 | |
DYDCUQKUCUHJBH-UWTATZPHSA-N | |
6234 | |
(4R)-4-amino-1,2-oxazolidin-3-one |
C3H6N2O2 | |
MFCD00005353 | |
d-cycloserine, cycloserine, seromycin, orientomycin, oxamycin, cyclorin, cyclo-d-serine, cicloserina, farmiserina, miroseryn | |
CHEBI:40009 | |
C1C(C(=O)NO1)N |
Specifications
68-41-7 | |
MFCD00005353 | |
DYDCUQKUCUHJBH-UWTATZPHSA-N | |
(4R)-4-amino-1,2-oxazolidin-3-one | |
6234 | |
25 mL | |
D-Cycloserine, 20mg/mL in distilled water, sterile-filtered |
C3H6N2O2 | |
d-cycloserine, cycloserine, seromycin, orientomycin, oxamycin, cyclorin, cyclo-d-serine, cicloserina, farmiserina, miroseryn | |
C1C(C(=O)NO1)N | |
102.093 | |
CHEBI:40009 | |
Liquid |
Safety and Handling
EINECSNumber : 200-688-4
RTECSNumber : NY2975000
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C