missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Galacto-D-mannan, Thermo Scientific Chemicals

$211.84 - $697.60

Chemical Identifiers

CAS 11078-30-1
MDL Number MFCD00146683
Synonym galactomannan, galactomannoglycan, d-galacto-d-mannane, 6-o-alpha-d-galactopyranosyl-4-o-beta-d-mannopyranosyl-beta-d-mannopyranose, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-2r,3s,4r,5s,6r-4,5,6-trihydroxy-3-2s,3s,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxyoxan-2-yl methoxy oxane-3,4,5-triol, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-2r,3s,4r,5s,6r-4,5,6-trihydroxy-3-2s,3s,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-2-yl methoxy tetrahydropyran-3,4,5-triol, 2r,3s,4s,5s,6s-2-hydroxymethyl-6-2r,3s,4r,5s,6r-4,5,6-trihydroxy-2-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-3-yl oxy oxane-3,4,5-triol
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66610MC
View Documents
Thermo Scientific Chemicals
J66610MC
100 mg
Each for $211.84
 
AAJ66610ME
View Documents
Thermo Scientific Chemicals
J66610ME
500 mg
Each for $697.60
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

11078-30-1
MFCD00146683
galactomannan, galactomannoglycan, d-galacto-d-mannane, 6-o-alpha-d-galactopyranosyl-4-o-beta-d-mannopyranosyl-beta-d-mannopyranose, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-2r,3s,4r,5s,6r-4,5,6-trihydroxy-3-2s,3s,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxyoxan-2-yl methoxy oxane-3,4,5-triol, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-2r,3s,4r,5s,6r-4,5,6-trihydroxy-3-2s,3s,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-2-yl methoxy tetrahydropyran-3,4,5-triol, 2r,3s,4s,5s,6s-2-hydroxymethyl-6-2r,3s,4r,5s,6r-4,5,6-trihydroxy-2-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-3-yl oxy oxane-3,4,5-triol

Specifications

11078-30-1
MFCD00146683
galactomannan, galactomannoglycan, d-galacto-d-mannane, 6-o-alpha-d-galactopyranosyl-4-o-beta-d-mannopyranosyl-beta-d-mannopyranose, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-2r,3s,4r,5s,6r-4,5,6-trihydroxy-3-2s,3s,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxyoxan-2-yl methoxy oxane-3,4,5-triol, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-2r,3s,4r,5s,6r-4,5,6-trihydroxy-3-2s,3s,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-2-yl methoxy tetrahydropyran-3,4,5-triol, 2r,3s,4s,5s,6s-2-hydroxymethyl-6-2r,3s,4r,5s,6r-4,5,6-trihydroxy-2-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-3-yl oxy oxane-3,4,5-triol
Powder
White
100 mg
∽200000
D-Galacto-D-mannan
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 234-299-6

RTECSNumber : OP3805000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only