missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Galacturonic acid monohydrate, 97%, Thermo Scientific Chemicals
$85.10 - $246.53
Chemical Identifiers
CAS | 91510-62-2 |
---|---|
Molecular Formula | C6H9O7 |
Molecular Weight (g/mol) | 0.00 |
MDL Number | MFCD00071585 |
InChI Key | AEMOLEFTQBMNLQ-DTEWXJGMSA-M |
Synonym | d-+-galacturonic acid monohydrate |
PubChem CID | 91872458 |
IUPAC Name | (2S,3R,4S,5R,6S)-3,4,5,6-tetrahydroxyoxane-2-carboxylic acid;hydrate |
SMILES | * |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6628206
|
Thermo Scientific Chemicals
J6628206 |
5 g |
Each for $85.10
|
|
|||||
AAJ6628214
|
Thermo Scientific Chemicals
J6628214 |
25 g |
Each for $246.53
|
|
|||||
Description
D-(+)-Galacturonic acid monohydrate is a chemical used in the synthesis of N-(D-galacturonoyl) amino acids and dipeptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-(+)-Galacturonic acid monohydrate is a chemical used in the synthesis of N-(D-galacturonoyl) amino acids and dipeptides.
Solubility
Soluble in water
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
91510-62-2 | |
0.00 | |
AEMOLEFTQBMNLQ-DTEWXJGMSA-M | |
91872458 | |
* |
C6H9O7 | |
MFCD00071585 | |
d-+-galacturonic acid monohydrate | |
(2S,3R,4S,5R,6S)-3,4,5,6-tetrahydroxyoxane-2-carboxylic acid;hydrate |
Specifications
91510-62-2 | |
C6H9O7 | |
5 g | |
14,4337 | |
Soluble in water | |
+53° (c=10 in Water) | |
(2S,3R,4S,5R,6S)-3,4,5,6-tetrahydroxyoxane-2-carboxylic acid;hydrate | |
91872458 | |
97% | |
D-Galacturonic acid monohydrate |
White | |
MFCD00071585 | |
1727087 | |
d-+-galacturonic acid monohydrate | |
AEMOLEFTQBMNLQ-DTEWXJGMSA-M | |
* | |
0.00 | |
212.15 | |
Powder |
Safety and Handling
EINECSNumber : 211-682-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only