missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals D-Glucosamine hydrochloride, 98+%

D-Glucosamine hydrochloride, CAS # 3416-24-8, is the salt of the amino sugar D-Glucosamine, used for chitin quantitation and other biochemical study.

Supplier:  Thermo Scientific Chemicals A1553230

 View more versions of this product

Catalog No. AAA1553230


Explore available promotions

Description

Description

Novel application of glucosamine is to prepare medical agent for treating vertigo. It is used as food ingredients and additives, raw material for anti cancer and antibiotic drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• D-Glucosamine is an abundant monosaccharide amino sugar that acts as a building block in the synthesis of glycosylated proteins and lipids

• D-Glucosamine is the precursor at the start of the hexosamine pathway that is essential to the formation of common proteoglycans

• D-Glucosamine combines with chondroitin sulfate in cartilage and is found as a monomer in chitin

Application

• D-Glucosamine can be used as a biochemical standard or for cell stimulation along with glucose in cell culture

• It may be useful in the laboratory study of osteoarthritis and diabetes

Specifications

Specifications

D-Glucosamine hydrochloride
∼192°C (decomposition)
MFCD00067674
4157370
Soluble in water.
215.64
66-84-2
C6H13NO5·HCl
250 g
14,4458
+71.5° (c=1 in Water, 20h)
≥98%

Chemical Identifiers

66-84-2
C6H13NO5·HCl
MFCD00067674
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-638-1

RTECSNumber : LZ6665000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only