missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-(+)-Glucose, anhydrous, For ACS analysis, MP Biomedicals™
$81.74 - $522.63
Chemical Identifiers
CAS | 50-99-7 |
---|---|
Molecular Formula | C6H12O6 |
Molecular Weight (g/mol) | 180.156 |
InChI Key | GZCGUPFRVQAUEE-SLPGGIOYSA-N |
Synonym | 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form |
PubChem CID | 107526 |
ChEBI | CHEBI:42758 |
IUPAC Name | (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal |
SMILES | C(C(C(C(C(C=O)O)O)O)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN15252790
|
MP Biomedicals Inc
0215252790 |
500 g |
Each for $81.74
|
|
|||||
ICN15252794
|
MP Biomedicals Inc
0215252794 |
5 kg |
Each for $522.63
|
|
|||||
Chemical Identifiers
50-99-7 | |
180.156 | |
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form | |
CHEBI:42758 | |
C(C(C(C(C(C=O)O)O)O)O)O |
C6H12O6 | |
GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
107526 | |
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal |
Specifications
50-99-7 | |
C6H12O6 | |
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form | |
C(C(C(C(C(C=O)O)O)O)O)O | |
180.156 | |
CHEBI:42758 | |
Powder |
1.544g/mL | |
500 g | |
GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal | |
107526 | |
180.2 | |
D-(+)-Glucose, For ACS analysis |