missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 388190010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP | |
146.0°C | |
C6H12O6 | |
15,4495 | |
+ 52.75 | |
Solubility in water: soluble. | |
C(C(C(C(C(C=O)O)O)O)O)O | |
180.16 | |
CHEBI:42758 | |
97.5 to 102.0% (Anhydrous substance) |
50-99-7 | |
0.5% max. (1 g, 105°C, 16 h) | |
1 kg | |
+ 52.75 (25.00°C c=10, H2O, NH4OH) | |
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form | |
GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal | |
107526 | |
180.16 | |
0.5% max. (total impurities) |
Chemical Identifiers
50-99-7 | |
180.16 | |
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form | |
CHEBI:42758 | |
C(C(C(C(C(C=O)O)O)O)O)O |
C6H12O6 | |
GZCGUPFRVQAUEE-SLPGGIOYSA-N | |
107526 | |
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal |
Safety and Handling
EINECSNumber : 200-075-1
RUO – Research Use Only