missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 388190050

 View more versions of this product

Catalog No. AC388190050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP
146.0°C
C6H12O6
15,4495
+ 52.75
Solubility in water: soluble.
C(C(C(C(C(C=O)O)O)O)O)O
180.16
CHEBI:42758
97.5 to 102.0% (Anhydrous substance)
50-99-7
0.5% max. (1 g, 105°C, 16 h)
5 kg
+ 52.75 (25.00°C c=10, H2O, NH4OH)
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
GZCGUPFRVQAUEE-SLPGGIOYSA-N
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
107526
180.16
0.5% max. (total impurities)

Chemical Identifiers

50-99-7
180.16
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
CHEBI:42758
C(C(C(C(C(C=O)O)O)O)O)O
C6H12O6
GZCGUPFRVQAUEE-SLPGGIOYSA-N
107526
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-075-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only