missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals D-(+)-Glucose monohydrate
$75.04 - $236.26
Chemical Identifiers
CAS | 14431-43-7 |
---|---|
Molecular Formula | C6H12O6·H2O |
MDL Number | MFCD00149450 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1109036
|
Thermo Scientific Chemicals
A1109036 |
500 g |
Each for $75.04
|
|
||||||
AAA110900E
|
Thermo Scientific Chemicals
A110900E |
2500 g |
Each for $236.26
|
|
||||||
Description
D-(+)-Glucose monohydrate is used by industry professionals to indicate the degree to which a substance contains water, and in food industry, pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-(+)-Glucose monohydrate is used by industry professionals to indicate the degree to which a substance contains water, and in food industry, pharmaceuticals.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
14431-43-7 | |
C6H12O6·H2O | |
MFCD00149450 |
Specifications
14431-43-7 | |
99% | |
MFCD00149450 | |
Soluble in water. | |
D-(+)-Glucose monohydrate |
∽83°C | |
C6H12O6·H2O | |
500 g | |
198.18 (180.16anhydrous) |
Safety and Handling
EINECSNumber : 200-075-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only