missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D(+)-Glucuronic acid, sodium salt, monohydrate, 99%, Thermo Scientific Chemicals
$351.33 - $351.33
Chemical Identifiers
CAS | 14984-34-0 |
---|---|
Molecular Formula | C6H9NaO7·H2O |
Molecular Weight (g/mol) | 234.14 |
MDL Number | MFCD00151051 |
InChI Key | QKHMTHNLNZGTSP-JSCKKFHOSA-N |
Synonym | sodium d-glucuronate |
PubChem CID | 87235405 |
IUPAC Name | sodium;(2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid |
SMILES | C(=O)C(C(C(C(C(=O)O)O)O)O)O.[Na] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC204571000
|
Thermo Scientific Chemicals
204571000 |
100 g | Plastic bottle |
Each for $351.33
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14984-34-0 | |
234.14 | |
QKHMTHNLNZGTSP-JSCKKFHOSA-N | |
87235405 | |
C(=O)C(C(C(C(C(=O)O)O)O)O)O.[Na] |
C6H9NaO7·H2O | |
MFCD00151051 | |
sodium d-glucuronate | |
sodium;(2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid |
Specifications
14984-34-0 | |
White | |
99% | |
C6H9NaO7·H2O | |
100 g | |
10, 4327 | |
+ 21 | |
QKHMTHNLNZGTSP-JSCKKFHOSA-N | |
sodium;(2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid | |
87235405 | |
99% | |
D(+)-Glucuronic acid, sodium salt, monohydrate |
134°C to 140°C | |
Authentic | |
Plastic bottle | |
MFCD00151051 | |
03, 886 | |
+21° (20°C c=2,H2O,after 4hrs.) | |
sodium d-glucuronate | |
C(=O)C(C(C(C(C(=O)O)O)O)O)O.[Na] | |
234.14 | |
234.14 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : 239-065-7
RUO – Research Use Only