missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Glutamic acid 5-tert-butyl ester, 95%
CAS: 45125-00-6 | C9H17NO4 | 203.238 g/mol
$64.44 - $787.33
Chemical Identifiers
| CAS | 45125-00-6 |
|---|---|
| Molecular Formula | C9H17NO4 |
| Molecular Weight (g/mol) | 203.238 |
| MDL Number | MFCD00153437 |
| InChI Key | OIOAKXPMBIZAHL-ZCFIWIBFSA-N |
| Synonym | h-d-glu otbu-oh, r-2-amino-5-tert-butoxy-5-oxopentanoic acid, d-glutamicacid, 5-1,1-dimethylethyl ester, 2r-2-amino-5-tert-butoxy-5-oxopentanoic acid, ambotzhaa1231, l-glutamicacid5-tert-butylester, r-2-amino-5-tert-butoxy-5-oxopentanoicacid |
| PubChem CID | 7016383 |
| IUPAC Name | (2R)-2-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid |
| SMILES | CC(C)(C)OC(=O)CCC(C(=O)O)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6346303
|
Thermo Scientific Chemicals
H6346303 |
1 g |
Each for $64.44
|
|
|||||
|
AAH6346306
|
Thermo Scientific Chemicals
H6346306 |
5 g |
Each for $220.66
|
|
|||||
|
AAH6346314
|
Thermo Scientific Chemicals
H6346314 |
25 g |
Each for $787.33
|
|
|||||
Description
It is employed as a intermediate for pharmaceutical and organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a intermediate for pharmaceutical and organic synthesis.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
| 45125-00-6 | |
| 203.238 | |
| OIOAKXPMBIZAHL-ZCFIWIBFSA-N | |
| 7016383 | |
| CC(C)(C)OC(=O)CCC(C(=O)O)N |
| C9H17NO4 | |
| MFCD00153437 | |
| h-d-glu otbu-oh, r-2-amino-5-tert-butoxy-5-oxopentanoic acid, d-glutamicacid, 5-1,1-dimethylethyl ester, 2r-2-amino-5-tert-butoxy-5-oxopentanoic acid, ambotzhaa1231, l-glutamicacid5-tert-butylester, r-2-amino-5-tert-butoxy-5-oxopentanoicacid | |
| (2R)-2-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid |
Specifications
| 45125-00-6 | |
| MFCD00153437 | |
| Slightly soluble in water. | |
| CC(C)(C)OC(=O)CCC(C(=O)O)N | |
| 1 g | |
| 7016383 | |
| 95% | |
| D-Glutamic acid 5-tert-butyl ester |
| C9H17NO4 | |
| h-d-glu otbu-oh, r-2-amino-5-tert-butoxy-5-oxopentanoic acid, d-glutamicacid, 5-1,1-dimethylethyl ester, 2r-2-amino-5-tert-butoxy-5-oxopentanoic acid, ambotzhaa1231, l-glutamicacid5-tert-butylester, r-2-amino-5-tert-butoxy-5-oxopentanoicacid | |
| OIOAKXPMBIZAHL-ZCFIWIBFSA-N | |
| (2R)-2-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid | |
| 203.238 | |
| 203.24 | |
| Solid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only