missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Histidine, 99%

CAS: 351-50-8 | C6H9N3O2 | 155.16 g/mol

Supplier:  Thermo Scientific Chemicals B2102714

 View more versions of this product

Catalog No. AAB2102714

Only null left
Explore available promotions

Description

Description

It is known to inhibit cell division, and is also used by certain types of bacteria (such as Escherichia coli) as a source of L-Histidine. It can be used in the design of peptide drugs, cationic peptides, such as analogues of carnosine and also be used as a heavy metal sequestration agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is known to inhibit cell division, and is also used by certain types of bacteria (such as Escherichia coli) as a source of L-Histidine. It can be used in the design of peptide drugs, cationic peptides, such as analogues of carnosine and also be used as a heavy metal sequestration agent.

Solubility
Soluble in water and 1 M HCl (50 mg/ml).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

∼285°C (decomposition)
351-50-8
MFCD00065963
d-histidine, h-d-his-oh, d-histidin, r-2-amino-3-1h-imidazol-4-yl propanoic acid, d-his, 2r-2-amino-3-1h-imidazol-4-yl propanoic acid, r-2-amino-3-4-imidazolyl propionic acid, 2r-2-amino-3-1h-imidazol-5-yl propanoic acid, r-alpha-amino-1h-imidazole-4-propionic acid, d-alpha-amino-beta-4-imidazolyl propionic acid
HNDVDQJCIGZPNO-RXMQYKEDSA-N
(2R)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
155.16
CHEBI:27947
155.16
D-Histidine
C6H9N3O2
84089
Soluble in water and 1 M HCl (50mg/ml).
N[C@H](CC1=CN=CN1)C(O)=O
25 g
71083
+39° (c=5 in Water)
99%

Chemical Identifiers

351-50-8
155.16
HNDVDQJCIGZPNO-RXMQYKEDSA-N
71083
(2R)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
C6H9N3O2
MFCD00065963
d-histidine, h-d-his-oh, d-histidin, r-2-amino-3-1h-imidazol-4-yl propanoic acid, d-his, 2r-2-amino-3-1h-imidazol-4-yl propanoic acid, r-2-amino-3-4-imidazolyl propionic acid, 2r-2-amino-3-1h-imidazol-5-yl propanoic acid, r-alpha-amino-1h-imidazole-4-propionic acid, d-alpha-amino-beta-4-imidazolyl propionic acid
CHEBI:27947
N[C@H](CC1=CN=CN1)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 206-513-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only