missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Histidine methyl ester dihydrochloride, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6509614
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
D-Histidine methyl ester dihydrochloride | |
White | |
MFCD00066137 | |
d-histidine methyl ester dihydrochloride, r-methyl 2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, h-d-his-ome 2hcl, h-d-his-ome.2hcl, d-histidine methylester dihydrochloride, c7h11n3o2.2hcl, methyl 2r-2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, d-histidine methyl ester di hydrochloride, d-his-ome.2hcl, d-histidinemethylesterdihydrochloride | |
COC(=O)C(CC1=CN=CN1)N.Cl.Cl | |
25 g | |
12658398 | |
Hygroscopic | |
Solid |
4467-54-3 | |
C7H13Cl2N3O2 | |
6491805 | |
DWAYENIPKPKKMV-QYCVXMPOSA-N | |
methyl (2R)-2-amino-3-(1H-imidazol-5-yl)propanoate;dihydrochloride | |
242.1 | |
242.1 | |
95% |
Chemical Identifiers
4467-54-3 | |
242.1 | |
DWAYENIPKPKKMV-QYCVXMPOSA-N | |
12658398 | |
COC(=O)C(CC1=CN=CN1)N.Cl.Cl |
C7H13Cl2N3O2 | |
MFCD00066137 | |
d-histidine methyl ester dihydrochloride, r-methyl 2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, h-d-his-ome 2hcl, h-d-his-ome.2hcl, d-histidine methylester dihydrochloride, c7h11n3o2.2hcl, methyl 2r-2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, d-histidine methyl ester di hydrochloride, d-his-ome.2hcl, d-histidinemethylesterdihydrochloride | |
methyl (2R)-2-amino-3-(1H-imidazol-5-yl)propanoate;dihydrochloride |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only