missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals D-Isoleucine, 98%
$496.01 - $1290.36
Chemical Identifiers
CAS | 319-78-8 |
---|---|
Molecular Formula | C6H13NO2 |
Molecular Weight (g/mol) | 131.18 |
MDL Number | MFCD00064221 |
InChI Key | AGPKZVBTJJNPAG-LXDRQGDMNA-N |
Synonym | d-isoleucine, 2r,3r-2-amino-3-methylpentanoic acid, h-d-ile-oh, isoleucine, d, unii-v87gja0g54, r-isoleucine, 2r,3r-2-amino-3-methylvaleric acid, alloisoleucin, d-ile, ambotzhaa1186 |
PubChem CID | 76551 |
ChEBI | CHEBI:27730 |
IUPAC Name | (2R,3R)-2-amino-3-methylpentanoic acid |
SMILES | CC[C@@H](C)[C@@H](N)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH27488MD
|
Thermo Scientific Chemicals
H27488MD |
250 mg |
Each for $496.01
|
|
|||||
AAH2748803
|
Thermo Scientific Chemicals
H2748803 |
1 g |
Each for $1,290.36
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
319-78-8 | |
131.18 | |
AGPKZVBTJJNPAG-LXDRQGDMNA-N | |
76551 | |
(2R,3R)-2-amino-3-methylpentanoic acid |
C6H13NO2 | |
MFCD00064221 | |
d-isoleucine, 2r,3r-2-amino-3-methylpentanoic acid, h-d-ile-oh, isoleucine, d, unii-v87gja0g54, r-isoleucine, 2r,3r-2-amino-3-methylvaleric acid, alloisoleucin, d-ile, ambotzhaa1186 | |
CHEBI:27730 | |
CC[C@@H](C)[C@@H](N)C(O)=O |
Specifications
265°C (sublimation) | |
C6H13NO2 | |
d-isoleucine, 2r,3r-2-amino-3-methylpentanoic acid, h-d-ile-oh, isoleucine, d, unii-v87gja0g54, r-isoleucine, 2r,3r-2-amino-3-methylvaleric acid, alloisoleucin, d-ile, ambotzhaa1186 | |
CC[C@@H](C)[C@@H](N)C(O)=O | |
250 mg | |
76551 | |
−40° (c=5 in 5N HCl) | |
98% |
319-78-8 | |
MFCD00064221 | |
AGPKZVBTJJNPAG-LXDRQGDMNA-N | |
(2R,3R)-2-amino-3-methylpentanoic acid | |
131.18 | |
CHEBI:27730 | |
131.18 | |
D-Isoleucine |
Safety and Handling
EINECSNumber : 206-269-2
RTECSNumber : NR4700000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only