missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Lactic acid, Thermo Scientific Chemicals

Useful chiral synthon, building block for depsipeptides

Supplier:  Thermo Scientific Chemicals J6187103

 View more versions of this product

Catalog No. AAJ6187103


Explore available promotions

Description

Description

D-(-)-Lactic acid is a useful biochemical for proteomics research. It is also used in the production of chiral drug and in the chemical synthesis. Lactic acid is used as a reagent in organic synthesis (in the manufacture of adhesives). It is used in the leather, textile, and tanning industries. It may be used as a plasticizer, a catalyst, or an acidifying agent. Lactic acid has even been used as a flavoring agent in the manufacture of tobacco products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-Lactic acid is a useful biochemical for proteomics research. It is also used in the production of chiral drug and in the chemical synthesis. Lactic acid is used as a reagent in organic synthesis (in the manufacture of adhesives). It is used in the leather, textile, and tanning industries. It may be used as a plasticizer, a catalyst, or an acidifying agent. Lactic acid has even been used as a flavoring agent in the manufacture of tobacco products.

Solubility
Soluble in water (50mg/ml)

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Keep away from strong oxidizing agents, strong bases.
Specifications

Specifications

D-Lactic acid
White
MFCD00068313
14,5335
Soluble in water (50mg/ml)
CC(C(=O)O)O
90.078
CHEBI:42111
1 g
Low Melting Solid
10326-41-7
C3H6O3
1720254
d-lactic acid, r-2-hydroxypropanoic acid, r-lactic acid, --lactic acid, 2r-2-hydroxypropanoic acid, d-lactate, r---lactic acid, r-lactate, propanoic acid, 2-hydroxy-, 2r, d-milchsaeure
JVTAAEKCZFNVCJ-UWTATZPHSA-N
(2R)-2-hydroxypropanoic acid
61503
90.08
−0.4° (c=8 in Water)

Chemical Identifiers

10326-41-7
90.078
JVTAAEKCZFNVCJ-UWTATZPHSA-N
61503
(2R)-2-hydroxypropanoic acid
C3H6O3
MFCD00068313
d-lactic acid, r-2-hydroxypropanoic acid, r-lactic acid, --lactic acid, 2r-2-hydroxypropanoic acid, d-lactate, r---lactic acid, r-lactate, propanoic acid, 2-hydroxy-, 2r, d-milchsaeure
CHEBI:42111
CC(C(=O)O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H315
Causes serious eye damage.
Causes skin irritation.

P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362

H315-H318

EINECSNumber : 233-713-2

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only