missing translation for 'onlineSavingsMsg'
Learn More

D-Lyxose, 99%

CAS: 1114-34-7 | C5H10O5 | 150.13 g/mol

$132.23 - $529.59

Chemical Identifiers

CAS 1114-34-7
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00064362
InChI Key PYMYPHUHKUWMLA-UOWFLXDJSA-N
Synonym 2s,3s,4r-2,3,4,5-tetrahydroxypentanal, d--lyxose, d-lyxo-pentose, unii-m795k6hjol, aldehydo-d-lyxose, m795k6hjol, dl-lyxose, +--lyxose, d-lyxose 9ci
PubChem CID 65550
ChEBI CHEBI:16789
IUPAC Name (2S,3S,4R)-2,3,4,5-tetrahydroxypentanal
SMILES C(C(C(C(C=O)O)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AAA1700706
View Documents
Thermo Scientific Chemicals
A1700706
5 g
Each for $132.23
Only null left
 
AAA1700714
View Documents
Thermo Scientific Chemicals
A1700714
25 g
Each for $529.59
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1114-34-7
150.13
PYMYPHUHKUWMLA-UOWFLXDJSA-N
65550
(2S,3S,4R)-2,3,4,5-tetrahydroxypentanal
C5H10O5
MFCD00064362
2s,3s,4r-2,3,4,5-tetrahydroxypentanal, d--lyxose, d-lyxo-pentose, unii-m795k6hjol, aldehydo-d-lyxose, m795k6hjol, dl-lyxose, +--lyxose, d-lyxose 9ci
CHEBI:16789
C(C(C(C(C=O)O)O)O)O

Specifications

1114-34-7
1.545
MFCD00064362
1723083
14,5641
PYMYPHUHKUWMLA-UOWFLXDJSA-N
(2S,3S,4R)-2,3,4,5-tetrahydroxypentanal
65550
150.13
D-Lyxose
109°C to 115°C
C5H10O5
5 g
Hygroscopic
2s,3s,4r-2,3,4,5-tetrahydroxypentanal, d--lyxose, d-lyxo-pentose, unii-m795k6hjol, aldehydo-d-lyxose, m795k6hjol, dl-lyxose, +--lyxose, d-lyxose 9ci
C(C(C(C(C=O)O)O)O)O
150.13
CHEBI:16789
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-212-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only