missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-(+)-Malic acid, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1168806

 View more versions of this product

Catalog No. AAA1168806


Explore available promotions

Description

Description

D-(+)-Malic acid used as an acidulant and flavoring agent, food additive. And it is also used in the place of the less sour citric acid in sour sweets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-Malic acid used as an acidulant and flavoring agent, food additive. And it is also used in the place of the less sour citric acid in sour sweets.

Solubility
Soluble in methanol, ethanol, acetone, ether.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Specifications

D- (+)-Malic acid
100°C to 104°C
C4H6O5
5 g
14,5707
Soluble in methanol,ethanol,acetone,ether.
+28° (c=5.5 in pyridine)
(2R)-2-hydroxybutanedioic acid
92824
134.09
636-61-3
1.6
MFCD00004245
1723540
d-+-malic acid, d-malic acid, d +-malic acid, r-2-hydroxysuccinic acid, r-malic acid, malic acid, d, 2r-2-hydroxybutanedioic acid, l +-malic acid, d-malate
BJEPYKJPYRNKOW-UHFFFAOYNA-N
OC(CC(O)=O)C(O)=O
134.09
CHEBI:30796
≥98%

Chemical Identifiers

636-61-3
134.09
BJEPYKJPYRNKOW-UHFFFAOYNA-N
92824
(2R)-2-hydroxybutanedioic acid
C4H6O5
MFCD00004245
d-+-malic acid, d-malic acid, d +-malic acid, r-2-hydroxysuccinic acid, r-malic acid, malic acid, d, 2r-2-hydroxybutanedioic acid, l +-malic acid, d-malate
CHEBI:30796
OC(CC(O)=O)C(O)=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c

H315-H318-H335

EINECSNumber : 211-262-2

RTECSNumber : ON7260000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only