missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D(+)-Malic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 153500010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
D(+)-Malic acid | |
636-61-3 | |
98% | |
HO2CCH2CH(OH)CO2H | |
03, 417 | |
d-+-malic acid, d-malic acid, d +-malic acid, r-2-hydroxysuccinic acid, r-malic acid, malic acid, d, 2r-2-hydroxybutanedioic acid, l +-malic acid, d-malate | |
BJEPYKJPYRNKOW-UHFFFAOYNA-N | |
(2R)-2-hydroxybutanedioic acid | |
92824 | |
134.09 | |
1 g | |
Glass bottle |
99.0°C to 104.0°C | |
White | |
C4H6O5 | |
MFCD00004245 | |
15, 5771 | |
Solubility in water: soluble. | |
OC(CC(O)=O)C(O)=O | |
134.09 | |
CHEBI:30796 | |
98% | |
Authentic | |
Crystalline Powder |
Chemical Identifiers
636-61-3 | |
134.09 | |
BJEPYKJPYRNKOW-UHFFFAOYNA-N | |
92824 | |
(2R)-2-hydroxybutanedioic acid |
C4H6O5 | |
MFCD00004245 | |
d-+-malic acid, d-malic acid, d +-malic acid, r-2-hydroxysuccinic acid, r-malic acid, malic acid, d, 2r-2-hydroxybutanedioic acid, l +-malic acid, d-malate | |
CHEBI:30796 | |
OC(CC(O)=O)C(O)=O |
Safety and Handling
GHS P Statement Causes serious eye irritation.
GHS P Statement Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Danger
RUO – Research Use Only