missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Pantothenic acid calcium salt, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1660922

 View more versions of this product

Catalog No. AAA1660922


Explore available promotions

Description

Description

Vitamin B-5D-Pantothenic acid calcium salt hydrate acts as a precursor in the biosynthesis of coenzyme A. It also involved in the catabolism of fatty acids, preparation of heme and the neurotransmitter acetylcholine. It is involved in the regulation of gene expression and in signal transduction. Further, it is used in sphingolipids, cholesterol and steroid hormones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vitamin B-5D-Pantothenic acid calcium salt hydrate acts as a precursor in the biosynthesis of coenzyme A. It also involved in the catabolism of fatty acids, preparation of heme and the neurotransmitter acetylcholine. It is involved in the regulation of gene expression and in signal transduction. Further, it is used in sphingolipids, cholesterol and steroid hormones.

Solubility
Soluble in water.

Notes
Hygroscopic. Air, light and moisture sensitive. Incompatible with strong oxidizing agents. Store in a cool place. Keep the container tightly closed.
Specifications

Specifications

∼138°C (decomposition)
137-08-6
MFCD00002766
14,7015
Soluble in water.
CC(C)(CO)C(C(=O)NCCC(=O)[O-])O.CC(C)(CO)C(C(=O)NCCC(=O)[O-])O.O.[Ca+2]
100 g
16211719
476.54
98%
D-Pantothenic acid calcium salt hydrate
C18H34CaN2O11
3769272
calcium 3-2,4-dihydroxy-3,3-dimethylbutanamido propanoate hydrate, d-pantothenic acid calcium salt hydrate, calcium hydrate dipantothenate, calcium bis 3-2,4-dihydroxy-3,3-dimethylbutanoyl amino propanoate hydrate
KUKRZUPDYACRAM-GXIDORRSSA-L
calcium;3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]propanoate;hydrate
494.551
+27° (c=5 in Water)
Hygroscopic

Chemical Identifiers

137-08-6
494.551
KUKRZUPDYACRAM-GXIDORRSSA-L
16211719
CC(C)(CO)C(C(=O)NCCC(=O)[O-])O.CC(C)(CO)C(C(=O)NCCC(=O)[O-])O.O.[Ca+2]
C18H34CaN2O11
MFCD00002766
calcium 3-2,4-dihydroxy-3,3-dimethylbutanamido propanoate hydrate, d-pantothenic acid calcium salt hydrate, calcium hydrate dipantothenate, calcium bis 3-2,4-dihydroxy-3,3-dimethylbutanoyl amino propanoate hydrate
calcium;3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]propanoate;hydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-278-9

RTECSNumber : RU4375000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only