missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Phenylalanine 98.0+%, TCI America™
Supplier: TCI America P01355G
Specifications
D-Phenylalanine | |
White | |
MFCD00004270 | |
COLNVLDHVKWLRT-MRVPVSSYSA-N | |
(2R)-2-amino-3-phenylpropanoic acid | |
165.19 | |
CHEBI:16998 | |
≥98.0% (T) |
673-06-3 | |
C9H11NO2 | |
d-phenylalanine, h-d-phe-oh, 2r-2-amino-3-phenylpropanoic acid, d-phe, sabiden, phenylalanine d-form, alanine, phenyl-, d, d-diphenylalanine, r-2-amino-3-phenylpropanoic acid, d-alpha-amino-beta-phenylpropionic acid | |
N[C@H](CC1=CC=CC=C1)C(O)=O | |
5 g | |
71567 | |
165.19 | |
Crystalline Powder |
Chemical Identifiers
673-06-3 | |
165.19 | |
COLNVLDHVKWLRT-MRVPVSSYSA-N | |
71567 | |
(2R)-2-amino-3-phenylpropanoic acid |
C9H11NO2 | |
MFCD00004270 | |
d-phenylalanine, h-d-phe-oh, 2r-2-amino-3-phenylpropanoic acid, d-phe, sabiden, phenylalanine d-form, alanine, phenyl-, d, d-diphenylalanine, r-2-amino-3-phenylpropanoic acid, d-alpha-amino-beta-phenylpropionic acid | |
CHEBI:16998 | |
N[C@H](CC1=CC=CC=C1)C(O)=O |
Safety and Handling
EINECSNumber : (9)-1282
RTECSNumber : AY7533000
TSCA : Yes