missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Phenylglycinamide, 99%

CAS: 6485-67-2 | C8H10N2O | 150.18 g/mol

$176.33 - $176.33

Chemical Identifiers

CAS 6485-67-2
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.18
MDL Number MFCD06799064
InChI Key KIYRSYYOVDHSPG-SSDOTTSWSA-N
Synonym r-2-amino-2-phenylacetamide, 2r-2-amino-2-phenylacetamide, d--phenylglycinamide, d-phenylglycinamide, unii-2ndy0ko51e, 2r-2-amino-2-phenylethanamide, 2ndy0ko51e, d-phenylglycine amide, d--phenylglycinamide,, d---phenylglycine amide
PubChem CID 729987
IUPAC Name (2R)-2-amino-2-phenylacetamide
SMILES C1=CC=C(C=C1)C(C(=O)N)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC346700050
SDS View Documents
Thermo Scientific Chemicals
346700050
5 g Glass bottle
Each for $176.33
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6485-67-2
150.18
KIYRSYYOVDHSPG-SSDOTTSWSA-N
729987
C1=CC=C(C=C1)C(C(=O)N)N
C8H10N2O
MFCD06799064
r-2-amino-2-phenylacetamide, 2r-2-amino-2-phenylacetamide, d--phenylglycinamide, d-phenylglycinamide, unii-2ndy0ko51e, 2r-2-amino-2-phenylethanamide, 2ndy0ko51e, d-phenylglycine amide, d--phenylglycinamide,, d---phenylglycine amide
(2R)-2-amino-2-phenylacetamide

Specifications

6485-67-2
White to Yellow
98.5% min. (GC)
C8H10N2O
MFCD06799064
−48.5° (20°C c=0.55,EtOH)
r-2-amino-2-phenylacetamide, 2r-2-amino-2-phenylacetamide, d--phenylglycinamide, d-phenylglycinamide, unii-2ndy0ko51e, 2r-2-amino-2-phenylethanamide, 2ndy0ko51e, d-phenylglycine amide, d--phenylglycinamide,, d---phenylglycine amide
KIYRSYYOVDHSPG-SSDOTTSWSA-N
(2R)-2-amino-2-phenylacetamide
729987
99%
D(-)-Phenylglycinamide, 99%
125.0°C to 129.0°C
Authentic
Glass bottle
C6H5CH(NH2)C(O)NH2
5 g
− 48.50
Solubility in water: soluble. Other solubilities: better soluble ph <6
C1=CC=C(C=C1)C(C(=O)N)N
150.18
150.18
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only