missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D(-)-Quinic acid, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 160500250

 View more versions of this product

Catalog No. AC160500250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

D(-)-Quinic acid
166.0°C to 168.0°C
0.1% max.
Authentic
Glass bottle
(HO)4C6H7CO2H
25 g
- 44.00 (20.00°C c=11.2,H2O)
quinic acid, d---quinic acid, --quinic acid, chinic acid, kinic acid, chinasaure, quinate, l-quinic acid, d-quinic acid, unii-058c04bgyi
AAWZDTNXLSGCEK-UHFFFAOYNA-N
(3R,5R)-1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid
6508
98+%
77-95-2
White to Yellow
2% max.
98+%
C7H12O6
MFCD00003864
15,8175
− 44.00
Solubility in water: 400g/L (20°C). Other solubilities: soluble in glacial acetic acid and alcohol
OC1CC(O)(CC(O)C1O)C(O)=O
192.17
192.17
Crystalline Powder

Chemical Identifiers

77-95-2
192.17
AAWZDTNXLSGCEK-UHFFFAOYNA-N
6508
OC1CC(O)(CC(O)C1O)C(O)=O
C7H12O6
MFCD00003864
quinic acid, d---quinic acid, --quinic acid, chinic acid, kinic acid, chinasaure, quinate, l-quinic acid, d-quinic acid, unii-058c04bgyi
(3R,5R)-1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS Signal Word: Warning

EINECSNumber : 201-072-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only