missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D(-)-Quinic acid, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 160501000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
D(-)-Quinic acid | |
166.0°C to 168.0°C | |
0.1% max. | |
Authentic | |
Glass bottle | |
(HO)4C6H7CO2H | |
100 g | |
- 44.00 (20.00°C c=11.2,H2O) | |
quinic acid, d---quinic acid, --quinic acid, chinic acid, kinic acid, chinasaure, quinate, l-quinic acid, d-quinic acid, unii-058c04bgyi | |
AAWZDTNXLSGCEK-UHFFFAOYNA-N | |
(3R,5R)-1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid | |
6508 | |
98+% |
77-95-2 | |
White to Yellow | |
2% max. | |
98+% | |
C7H12O6 | |
MFCD00003864 | |
15,8175 | |
− 44.00 | |
Solubility in water: 400g/L (20°C). Other solubilities: soluble in glacial acetic acid and alcohol | |
OC1CC(O)(CC(O)C1O)C(O)=O | |
192.17 | |
192.17 | |
Crystalline Powder |
Chemical Identifiers
77-95-2 | |
192.17 | |
AAWZDTNXLSGCEK-UHFFFAOYNA-N | |
6508 | |
OC1CC(O)(CC(O)C1O)C(O)=O |
C7H12O6 | |
MFCD00003864 | |
quinic acid, d---quinic acid, --quinic acid, chinic acid, kinic acid, chinasaure, quinate, l-quinic acid, d-quinic acid, unii-058c04bgyi | |
(3R,5R)-1,3,4,5-tetrahydroxycyclohexane-1-carboxylic acid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS Signal Word: Warning
EINECSNumber : 201-072-8
RUO – Research Use Only