missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-(+)-Raffinose pentahydrate, 98%, ≥98%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210279780
Specifications
D-(+)-Raffinose pentahydrate | |
17629-30-0 | |
1.465g/mL | |
C18H42O21 | |
d-+-raffinosepentahydrate | |
C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O | |
594.513 | |
594.5 | |
Crystalline Powder |
For HPLC analysis | |
White | |
0.98 | |
100 g | |
BITMAWRCWSHCRW-ORBWWOJFSA-N | |
(3S,4S,5R,6R)-2-[[(2R,3S,4S,5S)-6-[(2S,3S,4R,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate | |
134129414 | |
≥98% |
Chemical Identifiers
17629-30-0 | |
594.513 | |
d-+-raffinosepentahydrate | |
(3S,4S,5R,6R)-2-[[(2R,3S,4S,5S)-6-[(2S,3S,4R,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate |
C18H42O21 | |
BITMAWRCWSHCRW-ORBWWOJFSA-N | |
134129414 | |
C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O |