missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-(+)-Raffinose pentahydrate, 98%, ≥98%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210279790

 View more versions of this product

Catalog No. ICN10279790


Explore available promotions

Description

Description

Specifications

Specifications

D-(+)-Raffinose pentahydrate
17629-30-0
1.465g/mL
C18H42O21
d-+-raffinosepentahydrate
C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O
594.513
594.5
Crystalline Powder
For HPLC analysis
White
0.98
500 g
BITMAWRCWSHCRW-ORBWWOJFSA-N
(3S,4S,5R,6R)-2-[[(2R,3S,4S,5S)-6-[(2S,3S,4R,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate
134129414
≥98%

Chemical Identifiers

17629-30-0
594.513
d-+-raffinosepentahydrate
(3S,4S,5R,6R)-2-[[(2R,3S,4S,5S)-6-[(2S,3S,4R,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate
C18H42O21
BITMAWRCWSHCRW-ORBWWOJFSA-N
134129414
C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O
Videos
SDS
Documents

Documents

Promotions

Promotions