missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-(+)-Raffinose Pentahydrate, Thermo Scientific Chemicals
Supplier: Thermo Fisher Scientific J2106036
Specifications
D-(+)-Raffinose Pentahydrate | |
78°C to 82°C | |
C18H42O21 | |
500 g | |
d-+-raffinosepentahydrate | |
C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O | |
594.513 | |
594.51 |
17629-30-0 | |
Plastic bottle | |
MFCD00071590 | |
4285763 | |
BITMAWRCWSHCRW-ORBWWOJFSA-N | |
(3 S,4 S,5 R,6 R)-2-[[(2 R,3 S,4 S,5 S)-6-[(2 S,3 S,4 R,5 R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate | |
134129414 | |
Crystalline Powder |
Chemical Identifiers
17629-30-0 | |
594.513 | |
BITMAWRCWSHCRW-ORBWWOJFSA-N | |
134129414 | |
C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3(C(C(C(O3)CO)O)O)CO)O)O)O)O)O)O)O.O.O.O.O.O |
C18H42O21 | |
MFCD00071590 | |
d-+-raffinosepentahydrate | |
(3 S,4 S,5 R,6 R)-2-[[(2 R,3 S,4 S,5 S)-6-[(2 S,3 S,4 R,5 R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;pentahydrate |
Safety and Handling
EINECSNumber : 208-146-9
RTECSNumber : LZ5851200
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only