missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Serine methyl ester hydrochloride, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6368514

 View more versions of this product

Catalog No. AAH6368514


Explore available promotions

Description

Description

D-Serine methyl ester hydrochloride is used as a intermediate for organic synthesis, pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Serine methyl ester hydrochloride is used as a intermediate for organic synthesis, pharmaceutical and chemical research.

Solubility
Soluble in ethanol and methanol.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

163°C to 166°C (decomposition)
5874-57-7
MFCD00066121
Soluble in ethanol and methanol.
CC(N(C)S(=O)(=O)C1=CC=C(Br)C=C1)C1=NC2=CC=CC=C2C(=O)N1C1=CC=C(Br)C=C1
25 g
11446470
95%
D-Serine methyl ester hydrochloride
C23H19Br2N3O3S
d-serine methyl ester hydrochloride, h-d-ser-ome.hcl, r-methyl 2-amino-3-hydroxypropanoate hydrochloride, d-serine, methyl ester, hydrochloride, d-serine methyl ester hcl, methyl 2r-2-amino-3-hydroxypropanoate hydrochloride, d-ser-ome hcl, cycloserine impurity 2, h-d-ser-ome.hci
LYRWWUCOZBTOPL-UHFFFAOYNA-N
4-bromo-N-{1-[3-(4-bromophenyl)-4-oxo-3,4-dihydroquinazolin-2-yl]ethyl}-N-methylbenzene-1-sulfonamide
577.29
155.58
Solid

Chemical Identifiers

5874-57-7
577.29
LYRWWUCOZBTOPL-UHFFFAOYNA-N
11446470
C23H19Br2N3O3S
MFCD00066121
d-serine methyl ester hydrochloride, h-d-ser-ome.hcl, r-methyl 2-amino-3-hydroxypropanoate hydrochloride, d-serine, methyl ester, hydrochloride, d-serine methyl ester hcl, methyl 2r-2-amino-3-hydroxypropanoate hydrochloride, d-ser-ome hcl, cycloserine impurity 2, h-d-ser-ome.hci
CC(N(C)S(=O)(=O)C1=CC=C(Br)C=C1)C1=NC2=CC=CC=C2C(=O)N1C1=CC=C(Br)C=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P280-P305+P351+P338-P310

H318

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only