missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals D-Sorbitol, 98%

Supplier:  Thermo Scientific Chemicals 036404A3

 View more versions of this product

Catalog No. AA36404A3


Explore available promotions

Description

Description

D-sorbitol is added to food products as sweetener. It is also used in industrial and medical applications. Sorbitol often is applied in modern cosmetics as a humectant and thickener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

D-Sorbitol
98°C to 100°C
C6H14O6
2 kg
Hygroscopic
Soluble in water,methanol,acetone and DMF.
C(C(C(C(C(CO)O)O)O)O)O
182.172
CHEBI:17924
Powder
50-70-4
98%
MFCD00004708
1721899
d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol
FBPFZTCFMRRESA-JGWLITMVSA-N
(2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
5780
182.17

Chemical Identifiers

50-70-4
182.172
FBPFZTCFMRRESA-JGWLITMVSA-N
5780
(2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
C6H14O6
MFCD00004708
d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol
CHEBI:17924
C(C(C(C(C(CO)O)O)O)O)O
Safety and Handling

Safety and Handling

EINECSNumber : 200-061-5

RTECSNumber : LZ4290000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only