missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D-Sucrose (Molecular Biology), Fisher BioReagents

For the preparation of density gradients for DNA ultracentrifugation

Supplier:  Fisher BioReagents BP22010

 View more versions of this product

Catalog No. BP220-10


Explore available promotions

Description

Description

D-Sucrose is suitable for the preparation of density gradients used in purification of proteins and nucleic acids by ultracentrifugation.
Specifications

Specifications

Sucrose
57-50-1
White
0.01% max.
DNase-, RNase- and Protease-Free
Poly Pail
MFCD00006626
15, 9012
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
CZMRCDWAGMRECN-PWPRYFECNA-N
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
5988
342.29
Molecular Biology
D-Sucrose
190°C
6.5 to 7.5
0.03% max.
≥99.9%
C12H22O11
10 kg
+66 to +67° (+ or -)
0.005% max. Insoluble Matter
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
342.30
CHEBI:17992
≥99.9%
Solid

Chemical Identifiers

57-50-1
342.30
CZMRCDWAGMRECN-PWPRYFECNA-N
5988
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
C12H22O11
MFCD00006626
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
CHEBI:17992
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:1
Flammability:1
Instability:0

Recommended Storage : RT

SDS Documents

Documents

Promotions

Promotions