missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

D(-)-Tartaric Acid (unnatural Tartaric Acid) 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 137862500

 View more versions of this product

Catalog No. AC137862500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

D(-)-Tartaric acid
167°C to 171°C
White
C4H6O6
MFCD00004238
16,304
d-tartaric acid, d---tartaric acid, 2s,3s-2,3-dihydroxysuccinic acid, d--tartaric acid, d-threaric acid, --d-tartaric acid, 2s,3s-2,3-dihydroxybutanedioic acid, --tartaric acid, s,s-tartaric acid, 2s,3s---tartaric acid
OC(C(O)C(O)=O)C(O)=O
150.09
CHEBI:15672
≥98.5%
Authentic
Plastic bottle
99%
147-71-7
99%
HO2CCH(OH)CH(OH)CO2H
03, 520
15, 9203
FEWJPZIEWOKRBE-UHFFFAOYNA-N
(2S,3S)-2,3-dihydroxybutanedioic acid
439655
150.09
250 g
0.5% max. (1 g, 105°C, 3 hrs)
Crystals or Crystalline Powder

Chemical Identifiers

147-71-7
150.09
FEWJPZIEWOKRBE-UHFFFAOYNA-N
439655
OC(C(O)C(O)=O)C(O)=O
C4H6O6
MFCD00004238
d-tartaric acid, d---tartaric acid, 2s,3s-2,3-dihydroxysuccinic acid, d--tartaric acid, d-threaric acid, --d-tartaric acid, 2s,3s-2,3-dihydroxybutanedioic acid, --tartaric acid, s,s-tartaric acid, 2s,3s---tartaric acid
CHEBI:15672
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 205-695-6

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only