missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals D-Threonine, 98%

$75.37 - $75.37

Chemical Identifiers

CAS 632-20-2
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064269
InChI Key AYFVYJQAPQTCCC-STHAYSLISA-N
Synonym d-threonine, h-d-thr-oh, 2r,3s-2-amino-3-hydroxybutanoic acid, threonine, d, r-threonine, d-threonin, d-2-amino-3-hydroxybutyric acid, 2r,3s-2-amino-3-hydroxybutyric acid, d-thr, dth
PubChem CID 69435
ChEBI CHEBI:16398
IUPAC Name (2R,3S)-2-amino-3-hydroxybutanoic acid
SMILES C[C@H](O)[C@@H](N)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC346970050
SDS View Documents
Thermo Scientific Chemicals
346970050
5 g Glass Bottle
Each for $75.37
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

632-20-2
119.12
AYFVYJQAPQTCCC-STHAYSLISA-N
69435
(2R,3S)-2-amino-3-hydroxybutanoic acid
C4H9NO3
MFCD00064269
d-threonine, h-d-thr-oh, 2r,3s-2-amino-3-hydroxybutanoic acid, threonine, d, r-threonine, d-threonin, d-2-amino-3-hydroxybutyric acid, 2r,3s-2-amino-3-hydroxybutyric acid, d-thr, dth
CHEBI:16398
C[C@H](O)[C@@H](N)C(O)=O

Specifications

200ppm max.
1ppm max.
200ppm max.
Authentic
0.3% max.
(TLC) Passes Test
Glass Bottle
MFCD00064269
300ppm max.
0.1% max.
AYFVYJQAPQTCCC-STHAYSLISA-N
(2R,3S)-2-amino-3-hydroxybutanoic acid
119.12
69435
119.12
Crystalline Powder or Crystals
632-20-2
White
10ppm max.
10ppm max.
98%
C4H9NO3
CH3CH(OH)CH(NH)COH
+27° to +29° (20°C, 589nm) (c=6, water)
98% min. (c=5, H2O, 430nm) 1cm cell
d-threonine, h-d-thr-oh, 2r,3s-2-amino-3-hydroxybutanoic acid, threonine, d, r-threonine, d-threonin, d-2-amino-3-hydroxybutyric acid, 2r,3s-2-amino-3-hydroxybutyric acid, d-thr, dth
C[C@H](O)[C@@H](N)C(O)=O
+28° (20°C c=6,water)
5 g
CHEBI:16398
≥97.5%
D-Threonine
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only