missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chemical Identifiers
CAS | 6138-23-4 |
---|---|
Molecular Formula | C12H26O13 |
Molecular Weight (g/mol) | 378.33 |
MDL Number | MFCD00071594 |
InChI Key | DPVHGFAJLZWDOC-DJCWUSJTNA-N |
Synonym | trehalose dihydrate, d-+-trehalose dihydrate, a,a-trehalose, d +-trehalose dihydrate, unii-7yin7j07x4, alpha,alpha-trehalose dihydrate, d-trehalose dihydrate, mycose,1-o-alpha-d-glucopyranosyl-alpha-d-glucopyranoside, alp,alp.-trehalose, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-3,4,5-triol |
PubChem CID | 181978 |
IUPAC Name | (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol;dihydrate |
SMILES | O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
BP268710
|
Fisher BioReagents
BP268710 |
10 g | Poly Bottle |
Each for $166.28
Case of 6 Each for $997.68
|
|
||||
BP268725
|
Fisher BioReagents
BP268725 |
25 g | Poly Bottle |
Each for $116.58
Case of 6 Each for $699.48
|
|
||||
BP2687100
|
Fisher BioReagents
BP2687100 |
100 g | Poly Bottle |
Each for $249.03
Case of 6 Each for $1,494.18
|
|
||||
BP26871
|
Fisher BioReagents
BP26871 |
1 kg | Poly Bottle |
Each for $1,927.13
Case of 6 Each for $11,562.78
|
|
||||
Description
D-(+)-Trehalose is used in several biochemical techniques.Chemical Identifiers
6138-23-4 | |
378.33 | |
DPVHGFAJLZWDOC-DJCWUSJTNA-N | |
181978 | |
O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O |
C12H26O13 | |
MFCD00071594 | |
trehalose dihydrate, d-+-trehalose dihydrate, a,a-trehalose, d +-trehalose dihydrate, unii-7yin7j07x4, alpha,alpha-trehalose dihydrate, d-trehalose dihydrate, mycose,1-o-alpha-d-glucopyranosyl-alpha-d-glucopyranoside, alp,alp.-trehalose, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-3,4,5-triol | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol;dihydrate |
Specifications
6138-23-4 | |
White | |
Conforms to reference | |
Poly Bottle | |
MFCD00071594 | |
15, 9742 | |
DPVHGFAJLZWDOC-DJCWUSJTNA-N | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol;dihydrate | |
181978 | |
98% min. | |
D-(+)-Trehalose, Dihydrate |
97°C | |
8.5 to 10.5% | |
≥98 % | |
C12H26O13 | |
10 g | |
trehalose dihydrate, d-+-trehalose dihydrate, a,a-trehalose, d +-trehalose dihydrate, unii-7yin7j07x4, alpha,alpha-trehalose dihydrate, d-trehalose dihydrate, mycose,1-o-alpha-d-glucopyranosyl-alpha-d-glucopyranoside, alp,alp.-trehalose, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-3,4,5-triol | |
O.O.OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O | |
378.33 | |
378.33 | |
Powder or Solid |
Safety and Handling
WARNING!
Emergency Overview
Irritating to eyes and skin. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:2
Flammability:1
Instability:1
Recommended Storage : RT