missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D(+)-Tryptophan, 99%
CAS: 153-94-6 | C11H12N2O2 | 204.23 g/mol
$900.46 - $900.46
Chemical Identifiers
| CAS | 153-94-6 |
|---|---|
| Molecular Formula | C11H12N2O2 |
| Molecular Weight (g/mol) | 204.23 |
| MDL Number | MFCD00005647 |
| InChI Key | QIVBCDIJIAJPQS-SECBINFHSA-N |
| Synonym | d-tryptophan, h-d-trp-oh, r-tryptophan, d +-tryptophan, d-trytophane, d-tryptophane, tryptophan, d, 2r-2-amino-3-1h-indol-3-yl propanoic acid, +-tryptophan, h-d-typ-oh |
| PubChem CID | 9060 |
| ChEBI | CHEBI:16296 |
| IUPAC Name | (2R)-2-amino-3-(1H-indol-3-yl)propanoic acid |
| SMILES | N[C@H](CC1=CNC2=CC=CC=C12)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC168631000
|
Thermo Scientific Chemicals
168631000 |
100 g |
Each for $900.46
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 153-94-6 | |
| 204.23 | |
| QIVBCDIJIAJPQS-SECBINFHSA-N | |
| 9060 | |
| (2R)-2-amino-3-(1H-indol-3-yl)propanoic acid |
| C11H12N2O2 | |
| MFCD00005647 | |
| d-tryptophan, h-d-trp-oh, r-tryptophan, d +-tryptophan, d-trytophane, d-tryptophane, tryptophan, d, 2r-2-amino-3-1h-indol-3-yl propanoic acid, +-tryptophan, h-d-typ-oh | |
| CHEBI:16296 | |
| N[C@H](CC1=CNC2=CC=CC=C12)C(O)=O |
Specifications
| 282.0°C to 285.0°C | |
| White to Yellow | |
| 0.5% max. | |
| C11H12N2O2 | |
| MFCD00005647 | |
| d-tryptophan, h-d-trp-oh, r-tryptophan, d +-tryptophan, d-trytophane, d-tryptophane, tryptophan, d, 2r-2-amino-3-1h-indol-3-yl propanoic acid, +-tryptophan, h-d-typ-oh | |
| QIVBCDIJIAJPQS-SECBINFHSA-N | |
| (2R)-2-amino-3-(1H-indol-3-yl)propanoic acid | |
| 100 g | |
| 9060 | |
| 204.23 | |
| Powder |
| 153-94-6 | |
| Authentic | |
| 99% | |
| Glass Bottle | |
| + 31.50 | |
| Solubility in water: 11g/L (20°C). Other solubilities: soluble in alkali hydroxides, soluble in hot alcohol, insoluble in chloroform | |
| N[C@H](CC1=CNC2=CC=CC=C12)C(O)=O | |
| + 31.50 (24.00°C c=1,H2O) | |
| 204.23 | |
| CHEBI:16296 | |
| 99% | |
| D(+)-Tryptophan |
RUO – Research Use Only