missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
D-Valine methyleester hydrochloride, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 398720010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
D-Valine methyleester hydrochloride | |
99% | |
Authentic | |
MFCD00237309 | |
d-valine methyl ester hydrochloride, methyl d-valinate hydrochloride, h-d-val-ome.hcl, r-methyl 2-amino-3-methylbutanoate hydrochloride, d-valinemethyl ester hydrochloride, methyl 2r-2-amino-3-methylbutanoate hydrochloride, d-valine, methyl ester, hydrochloride, h-d-val-ome, methyl d-valinate hcl, h-d-val-ome hcl | |
[Cl].COC(=O)C(N)C(C)C | |
-13.5° to -17.5° (20°C, 589nm) (c=2, H2O) | |
11984198 | |
≥98.5% |
7146-15-8 | |
C6H13ClNO2 | |
(CH3)2CHCH(NH2)COOCH3·HCl | |
Glass bottle | |
RFMIEHOYULVMNZ-UHFFFAOYNA-N | |
methyl (2R)-2-amino-3-methylbutanoate;hydrochloride | |
166.63 | |
167.63 | |
1g |
Chemical Identifiers
7146-15-8 | |
166.63 | |
RFMIEHOYULVMNZ-UHFFFAOYNA-N | |
11984198 | |
[Cl].COC(=O)C(N)C(C)C |
C6H13ClNO2 | |
MFCD00237309 | |
d-valine methyl ester hydrochloride, methyl d-valinate hydrochloride, h-d-val-ome.hcl, r-methyl 2-amino-3-methylbutanoate hydrochloride, d-valinemethyl ester hydrochloride, methyl 2r-2-amino-3-methylbutanoate hydrochloride, d-valine, methyl ester, hydrochloride, h-d-val-ome, methyl d-valinate hcl, h-d-val-ome hcl | |
methyl (2R)-2-amino-3-methylbutanoate;hydrochloride |
RUO â Research Use Only