missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

d-Vitamin E Succinate, USP, 96-102%, Spectrum™ Chemical

V1035, 4345-03-3, C33H54O5

$325.20 - $26729.46

Chemical Identifiers

CAS 4345-03-3
Molecular Formula C33H54O5
Molecular Weight (g/mol) 530.79
InChI Key IELOKBJPULMYRW-NJQVLOCASA-N
IUPAC Name 4-oxo-4-{[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl]oxy}butanoic acid
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(OC(=O)CCC(O)=O)=C(C)C(C)=C2O1
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18602359
View Documents
Spectrum Chemical Mfg Cor
V103525GM
25 g Amber Glass Bottle
Each for $325.20
 
18602360
View Documents
Spectrum Chemical Mfg Cor
V1035100GM
100 g Amber Glass Bottle
Each for $1,040.20
 
18602361
View Documents
Spectrum Chemical Mfg Cor
V1035500GM
500 g Amber Glass Bottle
Each for $2,689.60
 
18602362
View Documents
Spectrum Chemical Mfg Cor
V10351KG
1 kg Amber Glass Bottle
Each for $4,498.94
 
18602363
View Documents
Spectrum Chemical Mfg Cor
V103525KGBL
25 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical d-Vitamin E Succinate, USP is a fat-soluble antioxidant. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

4345-03-3
530.79
4-oxo-4-{[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl]oxy}butanoic acid
C33H54O5
IELOKBJPULMYRW-NJQVLOCASA-N
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(OC(=O)CCC(O)=O)=C(C)C(C)=C2O1

Specifications

4345-03-3
Amber Glass Bottle
25 g
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(OC(=O)CCC(O)=O)=C(C)C(C)=C2O1
530.79
USP
100%
C33H54O5
IELOKBJPULMYRW-NJQVLOCASA-N
4-oxo-4-{[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-yl]oxy}butanoic acid
96 to 102%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'