missing translation for 'onlineSavingsMsg'
Learn More

Dacarbazine

CAS: 4342-03-4 | C6H10N6O | 182.19 g/mol

Supplier:  Thermo Scientific Chemicals J6102303

 View more versions of this product

Catalog No. AAJ6102303

Only null left
Explore available promotions

Description

Description

Used in treatment of malignant melanoma and sarcomas. Generates methyl adducts to purine bases in DNA

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Used in treatment of malignant melanoma and sarcomas
  • Generates methyl adducts to purine bases in DNA
Specifications

Specifications

Dacarbazine
210°C to 212°C
C6H10N6O
1 g
dacarbazine, biocarbazine, deticene, dtic-dome, biocarbazine r, decarbazine, dtic, icdt, dacarbazinum inn-latin, dacarbazino inn-spanish
OMJKFYKNWZZKTK-UXBLZVDNSA-N
(5E)-5-(dimethylaminohydrazinylidene)imidazole-4-carboxamide
5353562
Powder
3-4-4342
White to Yellow
MFCD00057167
14,2798
Slightly soluble in water,DMSO and ethanol /n
CN(C)NN=C1C(=NC=N1)C(=O)N
182.19
182.18

Chemical Identifiers

3-4-4342
MFCD00057167
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H340-H350-H332-H315-H319-H335
Toxic in contact with skin.
May cause genetic defects.
May cause cancer.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P332+P313-P362-P501c

H312+H332-H315-H319-H335-H350

EINECSNumber : 224-396-1

RTECSNumber : NI3950000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only