missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Dansyl Chloride, Calbiochem™,

Reagent used for fluorescent labeling of proteins, N-terminal amino acids, and amines

Supplier:  MilliporeSigma™ 3111551GM

Catalog No. 3111551GM


Explore available promotions

Description

Description

Specifications

Specifications

Dansyl Chloride
1 g
≥98%
MFCD00003985
dansyl chloride, 5-dimethylamino naphthalene-1-sulfonyl chloride, dns chloride, dansyl, 5-dimethylamino-1-naphthalenesulfonyl chloride, 1-naphthalenesulfonyl chloride, 5-dimethylamino, 1-dimethylaminonaphthalene-5-sulfonyl chloride, 1-chlorosulfonyl-5-dimethylaminonaphthalene, unii-qmu9166tj4, 5-dimethylaminonaphthalene-1-sulphonyl chloride
CN(C)C1=CC=CC2=C(C=CC=C12)S(Cl)(=O)=O
269.74
CHEBI:51907
605-65-2
by assay
C12H12ClNO2S
UN3261
XPDXVDYUQZHFPV-UHFFFAOYSA-N
5-(dimethylamino)naphthalene-1-sulfonyl chloride
11801
269.8

Chemical Identifiers

605-65-2
269.74
XPDXVDYUQZHFPV-UHFFFAOYSA-N
11801
5-(dimethylamino)naphthalene-1-sulfonyl chloride
C12H12ClNO2S
MFCD00003985
dansyl chloride, 5-dimethylamino naphthalene-1-sulfonyl chloride, dns chloride, dansyl, 5-dimethylamino-1-naphthalenesulfonyl chloride, 1-naphthalenesulfonyl chloride, 5-dimethylamino, 1-dimethylaminonaphthalene-5-sulfonyl chloride, 1-chlorosulfonyl-5-dimethylaminonaphthalene, unii-qmu9166tj4, 5-dimethylaminonaphthalene-1-sulphonyl chloride
CHEBI:51907
CN(C)C1=CC=CC2=C(C=CC=C12)S(Cl)(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 210-092-6

RTECSNumber : QK3688000

Recommended Storage : Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 4°C (39.2°F).

SDS
Documents

Documents

Promotions

Promotions