missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dantrolene sodium salt, Thermo Scientific Chemicals

Inhibits intracellular calcium release from the sarcoplasmic reticulum

$145.36 - $325.05

Chemical Identifiers

CAS 14663-23-1
Molecular Formula C14H12N4NaO5
Molecular Weight (g/mol) 339.263
MDL Number MFCD00079130
InChI Key XHONPQLVCHTEDY-YVCISUPJSA-N
PubChem CID 131674048
IUPAC Name molecular hydrogen;1-[(E)-[5-(4-nitrophenyl)furan-2-yl]methylideneamino]imidazolidine-2,4-dione;sodium
SMILES [HH].C1C(=O)NC(=O)N1N=CC2=CC=C(O2)C3=CC=C(C=C3)[N+](=O)[O-].[Na]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60887MC
View Documents
Thermo Scientific Chemicals
J60887MC
100 mg
Each for $145.36
 
AAJ60887MD
View Documents
Thermo Scientific Chemicals
J60887MD
250 mg
Each for $325.05
 
Description

Description

Inhibits intracellular calcium release from the sarcoplasmic reticulum

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14663-23-1
339.263
XHONPQLVCHTEDY-YVCISUPJSA-N
molecular hydrogen;1-[(E)-[5-(4-nitrophenyl)furan-2-yl]methylideneamino]imidazolidine-2,4-dione;sodium
C14H12N4NaO5
MFCD00079130
131674048
[HH].C1C(=O)NC(=O)N1N=CC2=CC=C(O2)C3=CC=C(C=C3)[N+](=O)[O-].[Na]

Specifications

14663-23-1
C14H12N4NaO5
100 mg
XHONPQLVCHTEDY-YVCISUPJSA-N
molecular hydrogen;1-[(E)-[5-(4-nitrophenyl)furan-2-yl]methylideneamino]imidazolidine-2,4-dione;sodium
131674048
Crystalline Powder
Orange to Yellow
MFCD00079130
14,2816
[HH].C1C(=O)NC(=O)N1N=CC2=CC=C(O2)C3=CC=C(C=C3)[N+](=O)[O-].[Na]
339.263
336.23
Dantrolene sodium salt
Videos
Safety and Handling

Safety and Handling

RTECSNumber : MU3875000

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only