3M™ DBI-SALA™ ExoFit NEX™ Arc Flash Harness
PVC coated aluminum back D-ring, Nomex™/Kevlar™ fiber webbing and comfort padding, with locking quick connect buckle leg straps.
Supplier:  3M™ 7100237731
Catalog No. 19-070-716
Explore available promotions
Description
Description
  • Arc flash rated
  • Nomex™/ Kevlar™ material provides shoulder, back and leg comfort padding
  • Tech-Lite™ stand-up PVC coated aluminum back D-ring
  • Duo-Lok™ quick connect chest and leg straps
  • Leather insulators
  • Nomex™/ Kevlar™ fiber webbing
  • Belt loops
  • Built-in lanyard keepers
  • Revolver™ vertical torso adjusters
  • Impact Indicator
  • Integrated flame resistant trauma straps
  • Built-in reflective materials
  • Protected labels, equipped with i-Safe™
  • Specifications
    Specifications
    Duo-Lok™ Quick Connect
    Utilities
    Back
    Zinc Plated Steel
    Nomex™, Dupont™ Kevlar™ aramid fiber
    7000 lb.
    ANSI A10.32, OSHA 1926.502, ANSI Z359.4, ANSI Z359.1, ASTM F887, OSHA 1910.66
    190 kg
    Large
    420 lb.
    4.5 lb.
    Zinc-plated Steel
    Shoulder, back, and leg padding, no body belt, with subpelvic strap