3M™ DBI-SALA™ ExoFit NEX™ Construction Style Positioning Harness
Manufactured using materials that won’t add unnecessary weight for ultimate comfort. Featuring aluminum back D-ring, belt with pad and side D-rings, locking quick connect buckle leg straps, and comfort padding.
Supplier:  3M™ 7100210165
 View more versions of this product
Catalog No. 19-070-735
Explore available promotions
Description
Description
  • Hybrid shoulder, back and leg comfort padding
  • Stand-up Tech-Lite™ aluminum back D-ring
  • Body belt/hip pad with Tech-Lite™ aluminum side D-rings
  • Duo-Lok™ quick connect chest and leg straps
  • Repel™ webbing
  • Protective shoulder caps
  • Built-in lanyard keepers
  • Revolver™ vertical torso adjusters
  • Impact Indicator
  • Integrated trauma straps
  • Built-in reflective materials
  • Protected labels, equipped with i-Safe™
  • Specifications
    Specifications
    Duo-Lok™ Quick Connect
    6.3 lb.
    Repel™ Polyester
    420 lb.
    190 kg
    Medium
    Back
    OSHA 1910.66, ANSI Z359.3, OSHA 1926.502, ANSI Z359.1