3M™ DBI-Sala™ ExoFit™ Vest-Style Harness
No-tangle X design dons easily
Supplier: 3M™ 1107981
Description
Offering exceptional comfort and protection, the ExoFit™ features a no-tangle “X” design with memory fit, so it dons easily and only has to be adjusted once.
- Made of soft, lightweight, high-performance materials for all-day wearability
- Features built-in shoulder, hip, and leg padding with Lycra™ edging for chafe-free mobility
- Aero-Spacer™ Dri-Lex™ lining wicks moisture away from the body for cool, dry comfort
- Three quick-connect buckles facilitate rapid donning and adjustment even with just one hand
- With back D-ring for connection to fall arrest system; convenient built-in belt; and coated, corrosion-resistant hardware
Compliance
Meets all applicable OSHA, ANSI™, and CSA requirements, including ANSI Z359.
Specifications
X-Large | |
Back | |
Meets all applicable OSHA, ANSI, and CSA requirements, including ANSIFor Use With (Application) Z359. |