3M™ DBI-SALA™ Delta™ Tower Climbing Harness
Back, front and side D-rings, belt with pad, seat sling with positioning D-rings, tongue buckle leg straps.
Supplier:  3M™ 7100207396
 View more versions of this product
Catalog No. 19-809-251
Explore available promotions
Description
Description
  • No-tangle design
  • Stand-up back D-ring
  • Front D-ring
  • Body belt/hip pad with side D-rings
  • Tool Pouch D-rings
  • Seat sling with positioning D-rings
Specifications
Specifications
Vest-Style Tower Climbing Harness
Medium
Vest
ANSI A10.32, ANSI Z359.1, OSHA 1926.502, ANSI Z359.4, OSHA 1910.66, ANSI Z359.3
7 lb.
Repel™ Polyester
Back, Front, Side, Suspension